Ubbelohde viscometers (DIN), Type No. 541 01, Capillary No. 0a, Constant K = 0.005, calibrated for automatic measurements, with fill ing and cleaning tube to connect to automatic cleaners AVS 24 and AVS 26 1 * 1 items
Catalog # 616-3379
Supplier: SI Analytics
Frequently Bought Together