Order Entry
Ireland
Orders LinkContactUsLinkComponent
Filter discs
76,2 mm, 1,59 mm
  42300.KS
Filter discs
  42300.KS
Filter discs
76,2 mm, 1,59 mm
1 * 1 items
These quartz discs are manufactured by fusing naturally occurring crystalline silica.
  • Ground and polished
  • Ø:
    76,2 mm
  • Seq:
    03
  • Thickness:
    1,59 mm