EK 100, SPRING CLAMP, FOR LABORATORY FLASKS UP TO DIA. 65 MM, FOR EASY AND SAFE FIXING OF FLASKS ON THE BOTTOM OF INSERT BASKETS OR UTENSIL HOLDERS WITH MESH SIZES UP TO 12,5 X 12,5 MM 1 * 1 items
Catalog # 142-1811
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together