Order Entry
Ireland
Orders LinkContactUsLinkComponent
Tin(II) fluoride ≥97.5%

Tin(II) fluoride ≥97.5%

Plastic bottle for solids, 100 g
Catalog # 11544.22
Corrosive
Irritant
Tin(II) fluoride ≥97.5%
Catalog # 11544.22
CAS Number:  7783-47-3
Tin(II) fluoride ≥97.5%
Tin(II) fluoride ≥97.5%

Plastic bottle for solids, 100 g

CAS Number:  7783-47-3
Price
79,70 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97.5% CAS No.: 7783-47-3 Molecular Formula: F2Sn Formula Weight: 156.69 Physical Form: Crystalline Density: 4.57 Appearance: White to off-white Odour: Odourless Melting Point: 219° Boiling Point: 850° Merck Reference: 14,8784 UN No.: 3260 MDL No.: MFCD00042540

Synonyms:  Tin difluoride
  • CAS Number:7783-47-3
  • Formula:SnF₂
  • MDL Number:MFCD00042540
  • Molecular Weight:156.71 g/mol
  • Merck Index:14,8784
  • Boiling Pt:850 °C
  • Melting Pt:219 °C
  • Density:4,57
  • Storage Temperature:Ambient
  • EINECS:231-999-3
  • ADR:8,II
  •  :Density: 4.57 g/cm³ (20 °C)
  • UN:3260
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.