Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
ISO 23696-1: Water Quality — Determination of Nitrate in Water Using Small-Scale Sealed Tubes — Part 1
• Method: This part specifies the nitrate determination using the Dimethyl phenol (DMP) reaction.
• Citation Forms: The primary citation form is NO₃-N, with NO₃⁻ as an alternative.
• Spectroquant® Cell Tests 1.00614, 1.14563, and 1.14764 align with this norm, employing the DMP method.
Documents
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure Merck Segment Brochure
Product Literature
Test Kits, Nitrate, Spectroquant®, Supelco®
Additional Product Details
Specifications
Specifications
- Chemical method:2,6-Dimethylphenol
- Pk:100 Tests
- Description:Nitrate test
- For unit:Prove, Nova 60, Picco
- Range:0,10 - 25,0 mg/L NO₃ - N, 0,4 - 110,7 mg/L NO₃
- Cat. No.:1.09713.0001
- M-Clarity quality level:MQ100



