Rheodyne Needle Port 1 * 1 items
Catalog # 554-2174
Supplier: IDEX RHEODYNE
Frequently Bought Together
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Accessories for pipette controller, BRAND accu-jet® S
Supplier: Brand
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
Cole Parmer® Dri-Block® BH-250 Heaters
Supplier: Cole-Parmer
Storage Boxes for 50 Vials
Supplier: Agilent