Order Entry
Ireland
Orders LinkContactUsLinkComponent
Accessories for chromatography systems
Accessories for chromatography systems
Tubing connector TC-3, 3× female, O.Ø 1,8 mm, PP, 1
Catalog # 19-0047-01
Supplier:   Cytiva
Accessories for chromatography systems
Catalog # 19-0047-01
Supplier:  Cytiva
CAS Number:  
Accessories for chromatography systems
Accessories for chromatography systems
Tubing connector TC-3, 3× female, O.Ø 1,8 mm, PP, 1
CAS Number:  
Price
131,00 €
Log in for availability
1 * 1 items
Specifications
Specifications
  • Pk:
    1
  • Description:
    Tubing connector TC-3, 3× female, O.Ø 1,8 mm, PP
Specifications