Order Entry
Ireland
ContactUsLinkComponent
Quartz glass capsule in application temperatures of up to 1050C (maximum 1600C), 10 ml volume, circular, low form, translucent, h igh chemical resistance and purity. Diameter 40mm, height 15mm. 1 * 1 items
Catalog # TSLA73015
Quartz glass capsule in application temperatures of up to 1050C (maximum 1600C), 10 ml volume, circular, low form, translucent, h igh chemical resistance and purity. Diameter 40mm, height 15mm. 1 * 1 items
Catalog # TSLA73015
Frequently Bought Together