Cryo writing set, Nunc™
Cryo writing set, Assorted
Catalog # 811-0009
Supplier: Thermo Fisher Scientific
Cryo writing set, Nunc™
Catalog # 811-0009
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
For labelling cryomaterials such as cryotubes, cryoboxes and cryolabels. Set contains one of each colour pen: black, blue, red and green.
- Extremely durable ink
- Fine tip
- No colour change at low temperatures
Specifications
- Colour:Assorted
- Description:Cryo writing set
- Cat. No.:811-0009
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact