Order Entry
Ireland
ContactUsLinkComponent
TITRONIC® universal. Module 1: Basic unit ready to use with 50 ml dosing unit TZ 3160 with tubes, screw threads, titration tip TZ 15 03, electrode-/titration tip holder TZ 3660, stand rod TZ 3665, manual control TZ 3680, 230 V 1 * 1 items
Catalog # 661-4438
Supplier:  SI Analytics
TITRONIC® universal. Module 1: Basic unit ready to use with 50 ml dosing unit TZ 3160 with tubes, screw threads, titration tip TZ 15 03, electrode-/titration tip holder TZ 3660, stand rod TZ 3665, manual control TZ 3680, 230 V 1 * 1 items
Catalog # 661-4438
Supplier:  SI Analytics
Frequently Bought Together