Order Entry
Ireland
ContactUsLinkComponent
Saffron alcoholic solution Gurr 1 * 500 ml
Catalog # 350934L
Supplier:  VWR Chemicals
undefined
Saffron alcoholic solution Gurr 1 * 500 ml
Catalog # 350934L
Supplier:  VWR Chemicals

Related Information

Frequently Bought Together