Order Entry
Ireland
ContactUsLinkComponent
Aniline sulphate GPR 1 * 50 g
Catalog # 272302U
Supplier:  VWR Chemicals
undefined
Aniline sulphate GPR 1 * 50 g
Catalog # 272302U
Supplier:  VWR Chemicals

Related Information

Frequently Bought Together