(1S)-(-)-Borneol 97%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (-)-Borneol, L(-)-Borneol, (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
- CAS Number:464-45-9
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00003759
- Molecular Weight:154.25 g/mol
- Merck Index:13,01328
- Boiling Pt:210 °C (760 mmHg)
- Melting Pt:204…208 °C
- Density:1,011 g/cm³ (20 °C)
- Flash Pt:65 °C
- Storage Temperature:Ambient
- EINECS:207-353-1
- UN:1312
- ADR:4.1,III
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:204 to 208 °C
- GC:≥96.0 %
- Specific optical rotation:-34.7° to -37.7° (20 °C, 589 nm) (c=5, C2H5OH)
Specifications
- Pack type:Plastic bottle
- Pk:100 g



