Hydroxyurea 98%
Glass bottle for solids, 25 g
: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- :127-07-1
- :CH₄N₂O₂
- :MFCD00007943
- :76.05 g/mol
- :13,04874
- :296 °C (1013 hPa)
- :142…146 °C (dec.)
- :Refrigerator
- :204-821-7
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:142 to 146 °C
- Titration Iodometric:≥97.5 % (on dried substance)
- Loss on drying:≤1 % (1 g, 60 °C) (< 0.7 kPa)
- Pack type:Glass bottle for solids
- Pk:25 g



