Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Hydroxyurea 98%

Glass bottle for solids, 5 g
Catalog # ACRO151680050
Health
Hydroxyurea 98%
Catalog # ACRO151680050
CAS Number:  127-07-1
Hydroxyurea 98%
Hydroxyurea 98%

Glass bottle for solids, 5 g

CAS Number:  127-07-1
Price
116,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…146 °C (dec.)
  • Storage Temperature:Refrigerator
  • EINECS:204-821-7

 

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    142 to 146 °C
  • Titration Iodometric:
    ≥97.5 % (on dried substance)
  • Loss on drying:
    ≤1 % (1 g, 60 °C) (< 0.7 kPa)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications