Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Phenol red, pure indicator

Glass bottle for solids, 25 g
Catalog # ACRO151430250
Irritant
Phenol red, pure indicator
Catalog # ACRO151430250
CAS Number:  143-74-8
Phenol red, pure indicator
Phenol red, pure indicator

Glass bottle for solids, 25 g

CAS Number:  143-74-8
Price
107,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:13,07329
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:205-609-7
Specifications

Specification Test Results

  • Appearance:
    Red Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in 95% ethanol) Clear soluble
  • UV Lambda max:
    555 to 561 nm (in buffer)
  • Specific extinction E (1%/1cm):
    ≥1700 (at lambda max., in buffer)
  • Visual transition interval:
    from pH 6.8 (yellow) to pH 8.2 (red)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications