Thiostrepton, Millipore®
10 g
Catalog # 598226-10
Supplier: Merck Millipore (Calbiochem)
Thiostrepton, Millipore®
Catalog # 598226-10
Supplier: Merck Millipore (Calbiochem)
CAS Number: 1393-48-2
Jump to:
Product Details & Documents
Thiostrepton is a thiazole-containing peptide antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit.
Thiostrepton inhibits the function of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome. The thiostrepton-resistant gene is also commonly used as a selective marker for recombinant DNA/plasmid technologies.
Synonyms: N-[(7R,8S)-2-Carboxy-7,8-dihydro-8-hydroxy-4-[(1S)-1-hydroxyethyl]-7-quinolinyl]-L-isoleucyl-L-alanyl-2,3-didehydroalanyl-L-alanyl-2-[(4aR,11S,14Z,18S,21S,28S,32aS)-4a-amino-21-[(1S,2R)-1,2-dihydroxy-1-methylpropyl]-14-ethylidene-3,4,4a,9,10,11,12,13,14,18,19,20,21,27,28,32a-hexadecahydro-11,28-bis[(1R)-1-hydroxyethyl]-9,12,19,26-tetraoxo-17H,26H-8,5:18,15:25,22:32,29-tetranitrilo-5H,15H-pyrido[3,2-m][1,11,17,24,4,7,20,27]tetrathiatetraazacyclotriacontin-2-yl]-4-thiazolecarbonyl-2,3-didehydroalanyl-2,3-didehydro-alaninamide,(1→528)-lactone
- CAS Number:1393-48-2
- Formula:C₇₂H₈₅N₁₉O₁₈S₅
- MDL Number:MFCD00135828
- Molecular Weight:1664.887 g/mol
Specifications
Specifications
- Pk:10 g



