Thiostrepton, Millipore®
1 g
Catalog # 598226-1
Supplier: Merck Millipore (Calbiochem)
Thiostrepton, Millipore®
Catalog # 598226-1
Supplier: Merck Millipore (Calbiochem)
CAS Number: 1393-48-2
Jump to:
Product Details & Documents
Thiostrepton is a thiazole-containing peptide antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit.
Thiostrepton inhibits the function of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome. The thiostrepton-resistant gene is also commonly used as a selective marker for recombinant DNA/plasmid technologies.
Synonyms: N-[(7R,8S)-2-Carboxy-7,8-dihydro-8-hydroxy-4-[(1S)-1-hydroxyethyl]-7-quinolinyl]-L-isoleucyl-L-alanyl-2,3-didehydroalanyl-L-alanyl-2-[(4aR,11S,14Z,18S,21S,28S,32aS)-4a-amino-21-[(1S,2R)-1,2-dihydroxy-1-methylpropyl]-14-ethylidene-3,4,4a,9,10,11,12,13,14,18,19,20,21,27,28,32a-hexadecahydro-11,28-bis[(1R)-1-hydroxyethyl]-9,12,19,26-tetraoxo-17H,26H-8,5:18,15:25,22:32,29-tetranitrilo-5H,15H-pyrido[3,2-m][1,11,17,24,4,7,20,27]tetrathiatetraazacyclotriacontin-2-yl]-4-thiazolecarbonyl-2,3-didehydroalanyl-2,3-didehydro-alaninamide,(1→528)-lactone
- CAS Number:1393-48-2
- Formula:C₇₂H₈₅N₁₉O₁₈S₅
- MDL Number:MFCD00135828
- Molecular Weight:1664.887 g/mol
Specifications
Specifications
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



