Order Entry
Ireland
ContactUsLinkComponent
IL-6, Human, Biotrak Assay, ELISA - 96 wells (Range: 10-400pg/ml. Sensitivity: <1pg/ml. Sample volume: 50ul. 1.5L wash buffer suppli ed. Calibrated to the NIBSC reference lot 88/514) 1 * 96 Assays
Catalog # RPN2754
Supplier:  Cytiva
IL-6, Human, Biotrak Assay, ELISA - 96 wells (Range: 10-400pg/ml. Sensitivity: <1pg/ml. Sample volume: 50ul. 1.5L wash buffer suppli ed. Calibrated to the NIBSC reference lot 88/514) 1 * 96 Assays
Catalog # RPN2754
Supplier:  Cytiva
Frequently Bought Together