Order Entry
Ireland
ContactUsLinkComponent
MMP-3, Human, Biotrak Assay, ELISA - 96 wells (Specific for total MMP-3: Recognizes proMMP-3, active MMP-3 and complexed MMP-3. No c ross-reactivity with MMP-1, -2, -9. Range: 3.75-120ng/ml. Sensitivity: 2.35ng/ml) 1 * 96 Assays
Catalog # RPN2613
Supplier:  Cytiva
MMP-3, Human, Biotrak Assay, ELISA - 96 wells (Specific for total MMP-3: Recognizes proMMP-3, active MMP-3 and complexed MMP-3. No c ross-reactivity with MMP-1, -2, -9. Range: 3.75-120ng/ml. Sensitivity: 2.35ng/ml) 1 * 96 Assays
Catalog # RPN2613
Supplier:  Cytiva
Frequently Bought Together