Order Entry
Ireland
Orders LinkContactUsLinkComponent
Benzophenone 99%, pure

Benzophenone 99%, pure

Plastic bottle for solids, 500 g
Catalog # ACRO105565000
Health
Benzophenone 99%, pure
Catalog # ACRO105565000
CAS Number:  119-61-9
Benzophenone 99%, pure
Benzophenone 99%, pure

Plastic bottle for solids, 500 g

CAS Number:  119-61-9
Price
53,50 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Diphenylmethanone, Diphenyl ketone
  • CAS Number:119-61-9
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:13,01099
  • Boiling Pt:305 °C (760 mmHg)
  • Melting Pt:47…49 °C
  • Density:1,085 g/cm³ (20 °C)
  • Flash Pt:143 °C
  • Storage Temperature:Ambient
  • EINECS:204-337-6
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    47 to 49 °C
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.