Order Entry
Ireland
Orders LinkContactUsLinkComponent
Copper(I) iodide ≥98%

Copper(I) iodide ≥98%

1 kg
Catalog # 11606.A1
Irritant
Copper(I) iodide ≥98%
Catalog # 11606.A1
CAS Number:  7681-65-4
Copper(I) iodide ≥98%
Copper(I) iodide ≥98%

1 kg

CAS Number:  7681-65-4
Price
322,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Purity: 98% CAS No.: 7681-65-4 Molecular Formula: CuI Formula Weight: 190.45 Physical Form: Powder Density: 5.63 Refractive Index: 2.346 Sensitivity: Light, Air and Moisture Sensitive Appearance: Pale brown Odour: Weak Melting Point: 605° Boiling Point: ca 1290° Solubility: Insoluble in water, dilute acids, alcohol. Decomposes by conc. acids. Soluble in NH3 solution, iodides, thiosulfates Merck Reference: 14,2662 UN No.: 3077 MDL No.: MFCD00010978

As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2

Synonyms:  Cuprous iodide
  • CAS Number:7681-65-4
  • Formula:CuI
  • MDL Number:MFCD00010978
  • Molecular Weight:190.45 g/mol
  • Merck Index:14,2662
  • Boiling Pt:∼1290 °C
  • Melting Pt:605 °C
  • Density:5,63
  • Storage Temperature:Ambient
  • EINECS:231-674-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.