Measuring cylinders, low form, class B, PP
250 ml, 5 ml, 5,0 ± ml
Catalog # BRND41648
Supplier: Brand
Jump to:
Product Details & Documents
PP, low form, Class B.
- Calibrated to contain (TC, In)
- Embossed scale
- PP with high clarity
Robust cylinders which on exposure to temperatures up to 80 °C will not cause permanently exceeded tolerance/error limits.
Documents
Brand Volumetric Measurement in the Laboratory
Product Literature
Specifications
- Capacity:250 ml
- Division:5 ml
- Tolerance:5,0 ± ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



