Order Entry
Ireland
Orders LinkContactUsLinkComponent
Measuring cylinders, low form, class B, PP

Measuring cylinders, low form, class B, PP

25 ml, 0,5  ml, 0,5 ± ml
Catalog # BRND41620
Supplier:   Brand
Measuring cylinders, low form, class B, PP
Catalog # BRND41620
Supplier:  Brand
Measuring cylinders, low form, class B, PP
Measuring cylinders, low form, class B, PP

25 ml, 0,5  ml, 0,5 ± ml

Price
36,90 €
Log in for availability
1 * 10 items
Product Details & Documents

PP, low form, Class B.

  • Calibrated to contain (TC, In)
  • Embossed scale
  • PP with high clarity

Robust cylinders which on exposure to temperatures up to 80 °C will not cause permanently exceeded tolerance/error limits.

Specifications
  • Capacity:
    25 ml
  • Division:
    0,5  ml
  • Tolerance:
    0,5 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.