Order Entry
Ireland
ContactUsLinkComponent
Pteridophytes (Ferns and Fern Allies). - 15 Microscope Slides - Primitive ferns (Psilophytatae) 1. Psilotum, rhizome showing protost ele t.s. 2. Psilotum, stem showing squamous leaves, aktinostele t.s. Clubmosses (Lycopodiatae) 3. Lycopodium, of stem 1 * 15 items
Catalog # LIED78800
Supplier:  LIEDER JOHANNES
Pteridophytes (Ferns and Fern Allies). - 15 Microscope Slides - Primitive ferns (Psilophytatae) 1. Psilotum, rhizome showing protost ele t.s. 2. Psilotum, stem showing squamous leaves, aktinostele t.s. Clubmosses (Lycopodiatae) 3. Lycopodium, of stem 1 * 15 items
Catalog # LIED78800
Supplier:  LIEDER JOHANNES
Frequently Bought Together