Order Entry
Ireland
ContactUsLinkComponent
As 9-hi-6,0,connecting cable with exposed cable ends for direct connection (high impedance) of ph electrodes to eco-/quadroline® mon itorscable length 6 m 1 * 1 items
Catalog # WTWO108507
Supplier:  WTW ONLINE
As 9-hi-6,0,connecting cable with exposed cable ends for direct connection (high impedance) of ph electrodes to eco-/quadroline® mon itorscable length 6 m 1 * 1 items
Catalog # WTWO108507
Supplier:  WTW ONLINE
Frequently Bought Together