Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Lactamide 98%

Irritant
New Product
Supplier:   Apollo Scientific
Lactamide 98%
Supplier:  Apollo Scientific
New Product
Certificates

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  2-Hydroxypropionamide, Lactamide
  • CAS Number:2043-43-8
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008017
  • Molecular Weight:89,094 g/mol
  • EINECS:218-049-3
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 22,60 €
    22,60 €
    1 * 1 g
       22,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 46,70 €
    46,70 €
    1 * 5 g
       46,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 970,00 €
    1 970,00 €
    1 * 500 g
       1 970,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 541,00 €
    541,00 €
    1 * 100 g
       541,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 141,00 €
    141,00 €
    1 * 25 g
       141,00 €

    Specifications:
    New Product
    New Product