Order Entry
Ireland
Orders LinkContactUsLinkComponent
Hydroxyurea ≥98% Ribonucleotide reductase inhibitor

Hydroxyurea ≥98% Ribonucleotide reductase inhibitor

Marketsource
 :   Abcam
Hydroxyurea ≥98% Ribonucleotide reductase inhibitor
 :  Abcam
Marketsource
 
Hydroxyurea ≥98% Ribonucleotide reductase inhibitor

 

 

Ribonucleotide reductase inhibitor. Various biological activities including anticancer and antiretroviral properties. Induces apoptosis in vitro.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 :  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  •  :127-07-1
  •  :76,055 g/mol
 

 

  • Constituents::
    98% Hydroxyurea
Product Family Options

 

  •  
     
  •    
       
    1 * 5 g
    1 * 5 g - 92,30 €
    92,30 €
    1 * 5 g
       92,30 €

     
    Marketsource
    Marketsource

ibitors/Activators

 

  •  
     
  •    
       
    1 * 25 g
    1 * 25 g - 303,00 €
    303,00 €
    1 * 25 g
       303,00 €

     
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.