Order Entry
Ireland
ContactUsLinkComponent
ProCare Med 40 Rinse Aid
ProCare Med 40 Rinse Aid
Catalog # MIEL11848250
Supplier:  MIELE
New Product
ProCare Med 40 Rinse Aid
Catalog # MIEL11848250
Supplier:  MIELE
New Product

Specifications

  • Capacity:
    1 L
  • Description:
    ProCare Med 40 rinse aid
  • Storage temperature:
    5...25 °C
  • pH:
    7,0 (at 20 °C)

Specifications

About this item

Rinse aid, neutral, ideal for rinsing instruments and utensils, biocompatible.

  • Significantly shorter drying times with reduced staining

    Clear and clean rinsing by reducing surface tension

    Easier dosing

    Reduces water mark streaking on goods

    Approved biocompatibility and does not interfere with sterilisation process

Dosed into the final rinse water in washerdisinfectors, ProCare Med 40 shortens drying times for reusable, invasive and non-invasive, medical devices like surgical and MIS instruments, and utensils.

Suitable for large chamber washer disinfectors for shortening drying time of beds and distribution trolleys.

pH-neutral with good material compatibility. Suitable for stainless steel, (anodised) aluminium, non-ferrous metals, PVC, rubber, plastic, materials, ceramics. For polysulfone (PSU) and polyphenylsulfone (PPSU), PVC, rubber, suitability tests should be done prior to use. Depending on water quality, dosing of 0,3 to 1,0 ml/L provides good results.

Certifications:

GHS/CLP compliant.