Order Entry
Ireland
ContactUsLinkComponent
Tork Xpress® Multifold Hand Towels
Tork Xpress® Multifold Hand Towels
Catalog # SCAA150290
Supplier:  Essity
New Product
undefined
Tork Xpress® Multifold Hand Towels
Catalog # SCAA150290
Supplier:  Essity
New Product
Specifications
  • For:
    Compatible with Tork H2 dispensers
  • Packed:
    21 packs, 226 paper hand towels each; 2 ply
  • Description:
    Universal multifold hand towels
  • L×W:
    85×196 mm (Folded size ); 255×196 mm (Unfolded size)
Specifications
About this item
Tork Xpress® multifold hand towel, an economic value for money - hand towel suitable for basic needs.
  • One-at-a-time dispensing for reduced consumption and increased hygiene
  • Universal quality
    Compatible with H2 dispensers
Certifications:
FSC Recycled credit, SA-COC-008266; EU Ecolabel, SE/004/001.