Order Entry
Ireland
ContactUsLinkComponent
VWR® Large Volume Cell Culture Plates
VWR® Large Volume Cell Culture Plates
  732-4428
New Product
undefined
VWR® Large Volume Cell Culture Plates
  732-4428
New Product

 

  • No. of wells:
    6
  • Sterile:
    +
  • Description:
    Culture plate with lid, irradiated
  • Well volume:
    50 ml

 

 

6-well, round well plates for suspension cell culture.

  • Round well provides gentle mixing

    Lower shearing force on cells

    Compatible with laboratory automation

    Virgin polystyrene construction

    Height: 88 mm

  • Barcoded

The round well design is ideal for prokaryotic and eukaryotic cell culture. The round well provides an optimal swirling motion with low shear stress, preventing cell damage during incubation. Perfect for delicate cell lines such as CHO and hybridomas.

 :

Plates are individually bagged with a lid.