Mouse Recombinant S100A8 (from E. coli)
Catalog # ABCAAB235971-100
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Seq:1286
- Storage conditions:−80 °C
- Biological activity:Standard
- Animal-free:N
- Protein synonyms:Caga|Mrp8|S100a8|Protein S100-A8|Calgranulin-A|Chemotactic cytokine CP-10|Leukocyte L1 complex light chain|Migration inhibitory factor-related protein 8|Pro-inflammatory S100 cytokine|S100 calcium-binding protein A8|MRP-8|p8
- Protein/peptide name:S100A8
- Purity:>90%
- Tested applications:SDS-PAGE
Specifications
About this item
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.