Mouse Recombinant AHSG (from Insect cells)
Catalog # ABCAAB201896-50
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Insect cells
- Species:Mouse
- Seq:1202
- Storage conditions:−80 °C
- Endotoxin content:<1 EU/µg
- Biological activity:Standard
- Animal-free:N
- Protein synonyms:Fetua|Alpha-2-HS-glycoprotein|Countertrypin|Fetuin-A
- Protein/peptide name:AHSG
- Purity:>90%
- Concentration:0,25 mg/ml
- Tested applications:SDS-PAGE
Specifications
About this item
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.