Mouse Recombinant PSMA (from Baculovirus (Insect cells))
Catalog # ABCAAB219283-50
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Seq:1202
- Storage conditions:Store at −20 °C
- Endotoxin content:<1 EU/µg
- Biological activity:Standard
- Form:Liquid
- Animal-free:N
- Protein synonyms:Mopsm|Naalad1|Folh1|Glutamate carboxypeptidase 2|Folate hydrolase 1|Folylpoly-gamma-glutamate carboxypeptidase|Glutamate carboxypeptidase II|Membrane glutamate carboxypeptidase|N-acetylated-alpha-linked acidic dipeptidase I|Prostate-specific membrane antigen homolog|Pteroylpoly-gamma-glutamate carboxypeptidase|FGCP|GCPII|mGCP|NAALADase I
- Protein/peptide name:PSMA
- Purity:>85%
- Concentration:0,5 mg/ml
- Formulation:pH: 7,4; Constituents: 90% PBS, 10% Glycerol (glycerin, glycerine)
- Shipping temperature:20
- Tested applications:SDS-PAGE
Specifications
About this item
Recombinant Mouse PSMA protein (His tag) is a Mouse Fragment protein, in the 45 to 752 aa range, expressed in Baculovirus infected insect, with >85% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.