Assembly Gloves rubipor XS 2001, 60276
: UVEX
The uvex rubipor XS is a lightweight, elastic safety glove with breathable NBR impregnation.
- Lightweight, elastic safety glove with stretch cotton material
- Suitable for operating touch displays
- Ergonomic fit
The combination of the very thin layer of impregnation and the stretch cotton material provides an extremely good level of breathability. Palm and fingertips are coated with breathable special NBR impregnation, providing good tactile feel through the flexible stretch cotton material with elastane. Free of harmful substances in line with OEKO-Tex® Standard 100.
- Catalog No:
- UVEX6027607
- UVEX6027608
- UVEX6027609
- UVEX6027605
- UVEX6027610
- UVEX6027606
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
Frequently Bought Together