Order Entry
Ireland
ContactUsLinkComponent
Anti-IL3RA/CD123 Mouse Monoclonal Antibody [clone: 6H6] PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Anti-IL3RA/CD123 Mouse Monoclonal Antibody [clone: 6H6] PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  ABCAAB157332-50
 :  Abcam
undefined
Anti-IL3RA/CD123 Mouse Monoclonal Antibody [clone: 6H6] PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  ABCAAB157332-50
 :  Abcam

 

  • Size:
    100 µg
  • Pack type:
    Vial

 

 

PerCP/Cy5.5® Mouse monoclonal [6H6] to IL3RA/CD123.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL3RA/CD123
Clonality: Monoclonal
Clone: 6H6
PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human