Anti-Ikaros Rabbit Monoclonal Antibody [clone: EPR25259-176]
: Abcam
Frequently Bought Together
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Media bottles, square, sterile, Nalgene®
: Thermo Fisher Scientific
Transfer pipettes
: RATIOLAB
Cell culture multiwell plates, CELLSTAR®
: Greiner Bio-One
Serological pipettes, Stripette™
: Corning
VWR®, Sample Containers, with Attached Lids
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Cell strainers, Falcon®
: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection