Order Entry
Ireland
ContactUsLinkComponent
Centrifuge tubes, conical, ASTM
Centrifuge tubes, conical, ASTM
Catalog # BIBB3450/02MP
Supplier:  SciLabware
undefined
Centrifuge tubes, conical, ASTM
Catalog # BIBB3450/02MP
Supplier:  SciLabware

Specifications

  • Capacity:
    100 ml
  • Length:
    200 mm
  • Pk:
    2

Specifications

About this item

PYREX®, borosilicate glass.

  • In accordance with standard methods of testing for water and sediment in petroleum products
Certifications: Complies with ASTM D96, D91, D-893 and D-1796