Gas dispersion tubes
Supplier: SciLabware
Certificates
About this item
These Pyrex® borosilicate glass tubes for the dispersion of gas in liquids with sintered thimble allow for the choice of porosities to achieve optimum gas bubble size.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion with relatively high resistance to temperature changes
Frequently Bought Together