Lithiumbis(trimethylsilyl)amide 0.9-1.1 M in hexane
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt
CAS Number:4039-32-1
:
Formula:C₆H₁₈LiNSi₂
MDL Number:MFCD00008261
:167.33 g/mol
Density:0,716
Flash Pt:–31 °C (–24 °F)
Storage Temperature:Ambient
EINECS:223-725-6
UN:2924
ADR:3,II
:Packaged under argon in resealable ChemSeal™ bottles.
:GHS02
:GHS08
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together