Capillary columns, OPTIMA® 1701
Medium polar GC phase.
- Special selectivity due to high cyanopropyl content
- Reference column for structure identification in combination with column of polarity 5 (e.g., OPTIMA® 5)
- Maximum temperature for isothermal operation 280 °C
- Maximum temperature for short isotherms in a temperature program 300 °C
14% cyanopropyl-phenyl – 86% dimethylpolysiloxane phase.
USP code: G46.
Frequently Bought Together