Disinfectant tablets, PRESEPT™
Catalog # SPR25
Supplier: WESTERN LABORATORY SERVICE
Specifications
- Description:PRESEPT™ disinfectant tablets 100 x 2,5 gram
- Pk:100
Specifications
About this item
These 2,5 g tablets contain sodium dichloroisocyanurate (NaDCC) in an effervescent base. The aqueous solution is rapid acting, and is effective against bacteria, spores, fungi, algae, protozoa and viruses, (including AIDS virus) and is resistant to inactivation by organic soilage.
- Activity across the complete biocidal spectrum
- Suitable for cleaning and disinfecting work surfaces, utensils and linen etc
- Convenient tablet format, more stable and better biocidal activity than hypochlorites
- Solution is compatible with anionic and non ionic detergents