Capillary columns, OPTIMA® 5
Nonpolar GC phase.
- Standard phase with large range of applications
- Thick film columns ≥3 μm film are expecially recommended for analytes with higher concentration
Columns with 0,10 to 0,32 mm ID and films <3 μm: max. temperature for isothermal operation 340 °C, max. temperature for short isotherms in a temperature program 360 °C; 0,53 mm ID columns with films <3 μm: max. temperatures 320 and 340 °C, resp.; thick film columns with films >3 μm: max. temperatures 300 and 320 °C, resp.
5% phenyl – 95% methylpolysiloxane phase.
USP code: G27/G36.
Frequently Bought Together