Pipette tips, low retention, Premier
GREI739209BE
- Sterile:-
- Volume:0,5 - 250 µl
- Tip style:Graduated, low retention
- For use with:Gilson® and similar pipettors
- Pk:10.000
Premier low retention tips provide the ultimate pipetting precision and accuracy with the minimum of liquid carryover.
- Free of detectable DNase, RNase, human DNA and pyrogens
- Reference lines moulded at 10 µl, 50 µl and 100 µl provide a constant visual check
- Ideal for use with Gilson® and similar pipettors
Low retention pipette tips are manufactured from specific polymers using the latest mould construction and maintenance technology to provide high polish cores and cavities. These in turn, produce finished product with mirror-like internal and external surfaces. The resultant pipette tips are extremely hydrophobic ensuring minimum liquid retention both on the internal and external walls.