2,4,5,6-Tetraaminopyrimidine sulphate
Catalog # MOLE19915065-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:238.23 g/mol
EINECS:226-393-0
CAS Number:5392-28-9
Formula:C₄H₈N₆·H₂SO₄
MDL Number:MFCD00012786