3-Fluoro-DL-phenylalanine for synthesis
Catalog # 8.41409.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:183.2 g/mol
EINECS:220-104-1
UN:2811
ADR:6.1,II
CAS Number:2629-54-1
:456-88-2
Formula:C₉H₁₀FNO₂
MDL Number:MFCD00004273