Diethyl cyclobutane-1,1-dicarboxylate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3779-29-1
:
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00019261
Boiling Pt:96…98 °C (8 mmHg)
Density:1,050
Flash Pt:89 °C (192 °F)
:200.24 g/mol
Storage Temperature:Ambient
EINECS:223-239-4
Frequently Bought Together