Order Entry
Ireland
ContactUsLinkComponent
HPLC columns, Acclaim™ HILIC-10
HPLC columns, Acclaim™ HILIC-10
Certificates
undefined
HPLC columns, Acclaim™ HILIC-10
Specifications
  • Catalog No:
  • 555-7493
  • 555-7496
  • Max. pressure:
  • 8500 psi
  • 8500 psi
  • pH range:
  • 2,0 - 8,0
  • 2,0 - 8,0
  • Temperature limit:
  • 80 °C
  • 80 °C