n-Butyl phenyl ether ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1126-79-0
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00009438
:150.22 g/mol
Boiling Pt:210…211 °C
Melting Pt:–19 °C
Density:0,938
Flash Pt:82 °C (179 °F)
Storage Temperature:Ambient
EINECS:214-426-1
Related Information
Frequently Bought Together