Order Entry
Ireland
Orders LinkContactUsLinkComponent
4-Chlorophenyl phenyl ether

4-Chlorophenyl phenyl ether

Glass bottle for solids, 100 mg
Catalog # PROOCILULM24210.1
4-Chlorophenyl phenyl ether
Catalog # PROOCILULM24210.1
CAS Number:  7005-72-3
4-Chlorophenyl phenyl ether

Glass bottle for solids, 100 mg

CAS Number:  7005-72-3
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  4-Chlorodiphenyl ether, 1-Chloro-4-phenoxybenzene
  • CAS Number:7005-72-3
  • Formula:C₁₂H₉ClO
  • MDL Number:MFCD00055431
  • Molecular Weight:204.66 g/mol
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:–8 °C
  • Density:1,193 g/cm³ (20 °C)
  •  :Density: 1.193 g/cm³ (20 °C)
  • EINECS:230-281-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.